This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc01056778_m1
See other PPM1A GE Assays ›
Gene Symbol:
PPM1A
Gene Name:
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases:
PP2Calpha
Chromosome Location:
Chr.20: 7849403 - 7904654 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc01056778_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPM1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082698.1 NP_001076167.1 1 - 2 849 81
XM_008271817.2 XP_008270039.1 2 - 3 912 81
XM_008271818.2 XP_008270040.1 2 - 3 909 81
XM_008271819.2 XP_008270041.1 2 - 3 951 81
XM_008271820.2 XP_008270042.1 2 - 3 951 81
XM_017349342.1 XP_017204831.1 3 - 4 1074 81
GenBank mRNA S87757.1 - 1 - 2 849 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0