This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc03397347_m1
See other VCAM1 GE Assays ›
Gene Symbol:
VCAM1
Gene Name:
vascular cell adhesion molecule 1
Gene Aliases:
VCAM-1
Chromosome Location:
Chr.13: 63440580 - 63462573 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc03397347_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VCAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082152.1 NP_001075621.1 6 - 7 1634 67
XM_008263958.2 XP_008262180.1 6 - 7 1860 67
XM_008263959.2 XP_008262181.1 5 - 6 1293 67
GenBank mRNA AY212510.1 - 6 - 7 1634 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Oryctolagus cuniculus Inventoried Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0