This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc03398515_m1
See other PPP1R2 GE Assays ›
Gene Symbol:
PPP1R2
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 2
Gene Aliases:
IPP-2
Chromosome Location:
Chr.14: 91192553 - 91215560 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc03398515_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082744.2 NP_001076213.1 2 - 3 497 104
XM_017346649.1 XP_017202138.1 2 - 3 403 104
GenBank mRNA L20858.1 - 2 - 3 495 104
M95581.1 - 2 - 3 232 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase inhibitor phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0