This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc03398801_m1
See other PTPA GE Assays ›
Gene Symbol:
PTPA
Gene Name:
protein phosphatase 2 phosphatase activator
Gene Aliases:
PPP2R4, PTPRA
Chromosome Location:
Chr.Unknown: 1202 - 28306 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc03398801_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082680.1 NP_001076149.1 643 63
GenBank mRNA X73479.1 - 643 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase activator phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0