Assay Details
| Gene Symbol: RS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109823.1 | NP_001103293.1 | 5 - 6 | 519 | 69 | |
| GenBank mRNA | EU074985.1 | - | 5 - 6 | 519 | 69 | |
More Information
Set Membership: |
|

RS1
Genomic Map