Assay Details
| Gene Symbol: ADORA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082058.2 | NP_001075527.1 | 1 | 250 | 101 | |
| GenBank mRNA | AF145438.1 | - | 1 | 251 | 101 | |
| U90718.1 | - | 1 | 51 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001082058.2 transcripts.
Set Membership: |
|

ADORA3
Genomic Map