This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc03824881_g1
See other EEF1G GE Assays ›
Gene Symbol:
EEF1G EEF1G
Gene Name:
eukaryotic translation elongation factor 1 gamma
eukaryotic translation elongation factor 1 gamma
Gene Aliases:
-
-
Chromosome Location:
Chr.14: 45707623 - 45709022 on Build OryCun2.0
Chromosome Location:
Chr.Unknown: 1747558 - 1759063 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
-
Assay ID Oc03824881_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101703.1 NP_001095173.1 1 553 94
GenBank mRNA X68142.1 - 553 94
Gene Symbol: EEF1G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008274377.2 XP_008272599.1 1 636 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Oryctolagus cuniculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0