This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06676473_g1
See other YTHDF3 GE Assays ›
Gene Symbol:
YTHDF3
Gene Name:
YTH N6-methyladenosine RNA binding protein 3
Gene Aliases:
-
Chromosome Location:
Chr.3: 78938188 - 78984446 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06676473_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YTHDF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008255566.2 XP_008253788.1 6 - 7 2284 88
XM_008255568.2 XP_008253790.1 4 - 5 2070 88
XM_008255569.2 XP_008253791.1 5 - 6 2137 88
XM_008255570.2 XP_008253792.1 4 - 5 1979 88
XM_008255571.2 XP_008253793.1 4 - 5 1760 88
XM_017341338.1 XP_017196827.1 6 - 7 2301 88
XM_017341339.1 XP_017196828.1 5 - 6 2233 88
XM_017341340.1 XP_017196829.1 6 - 7 2211 88
XM_017341341.1 XP_017196830.1 5 - 6 2143 88
XM_017341342.1 XP_017196831.1 4 - 5 2086 88
XM_017341344.1 XP_017196833.1 2 - 3 1619 88
XM_017341345.1 XP_017196834.1 4 - 5 1911 88
XM_017341346.1 XP_017196835.1 4 - 5 1744 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0