This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06676772_m1
See other POLR3GL GE Assays ›
Gene Symbol:
POLR3GL
Gene Name:
RNA polymerase III subunit GL
Gene Aliases:
-
Chromosome Location:
Chr.13: 42753467 - 42768760 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06676772_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR3GL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008264531.2 XP_008262753.1 9 - 10 1013 71
XM_008264532.2 XP_008262754.1 9 - 10 1056 71
XM_008264537.2 XP_008262759.1 7 - 8 758 71
XM_017345927.1 XP_017201416.1 9 - 10 1036 71
XM_017345928.1 XP_017201417.1 9 - 10 1055 71
XM_017345929.1 XP_017201418.1 8 - 9 1020 71
XM_017345930.1 XP_017201419.1 9 - 10 891 71
XM_017345931.1 XP_017201420.1 8 - 9 998 71
XM_017345932.1 XP_017201421.1 7 - 8 962 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0