This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06677443_m1
See other NDNF GE Assays ›
Gene Symbol:
NDNF
Gene Name:
neuron derived neurotrophic factor
Gene Aliases:
-
Chromosome Location:
Chr.15: 96195537 - 96238199 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06677443_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDNF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002717229.3 XP_002717275.2 2 - 3 251 89
XM_008267904.2 XP_008266126.1 3 - 4 832 89
XM_008267905.2 XP_008266127.1 2 - 3 745 89
XM_008267906.2 XP_008266128.1 3 - 4 813 89
XM_017347551.1 XP_017203040.1 3 - 4 342 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0