Assay Details
| Gene Symbol: ASIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711142.2 | XP_002711188.1 | 2 - 3 | 465 | 58 | |
| XM_008256435.2 | XP_008254657.1 | 2 - 3 | 469 | 58 | ||
More Information
Set Membership: |
|

ASIC1
Genomic Map