This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06678453_m1
See other RBBP5 GE Assays ›
Gene Symbol:
RBBP5
Gene Name:
RB binding protein 5, histone lysine methyltransferase complex subunit
Gene Aliases:
-
Chromosome Location:
Chr.16: 67011594 - 67049827 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06678453_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBBP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002717447.3 XP_002717493.1 2 - 3 186 65
XM_008268525.2 XP_008266747.1 2 - 3 187 65
XM_008268526.2 XP_008266748.1 2 - 3 187 65
XM_008268527.2 XP_008266749.1 2 - 3 187 65
XM_008268528.2 XP_008266750.1 2 - 3 188 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5lprg:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0