Assay Details
| Gene Symbol: LOC100358333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017341006.1 | XP_017196495.1 | 2 - 3 | 1398 | 93 | |
| XM_017341007.1 | XP_017196496.1 | 2 - 3 | 1398 | 93 | ||
| XM_017341008.1 | XP_017196497.1 | 2 - 3 | 1398 | 93 | ||
More Information
Set Membership: |
|

LOC100358333
Genomic Map