Assay Details
| Gene Symbol: EPCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709724.3 | XP_002709770.1 | 8 - 9 | 1118 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPCAM| Gene Symbol: EPCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709724.3 | XP_002709770.1 | 8 - 9 | 1118 | 75 | |
Set Membership: |
|