This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06708853_m1
See other SLCO1A2 GE Assays ›
Gene Symbol:
SLCO1A2
Gene Name:
solute carrier organic anion transporter family member 1A2
Gene Aliases:
OATP1
Chromosome Location:
Chr.8: 18713326 - 18818550 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06708853_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO1A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008259561.2 XP_008257783.1 12 - 13 1825 63
XM_008259562.2 XP_008257784.1 12 - 13 1788 63
XM_017343348.1 XP_017198837.1 12 - 13 1856 63
XM_017343349.1 XP_017198838.1 12 - 13 1853 63
XM_017343350.1 XP_017198839.1 12 - 13 1783 63
GenBank mRNA KC708581.1 - 9 - 10 1310 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0