This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06709307_m1
See other ASXL2 GE Assays ›
Gene Symbol:
ASXL2
Gene Name:
ASXL transcriptional regulator 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 159968804 - 160145591 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06709307_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASXL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008254661.2 XP_008252883.1 11 - 12 2165 59
XM_008254662.2 XP_008252884.1 11 - 12 2163 59
XM_008254663.2 XP_008252885.1 11 - 12 2165 59
XM_008254664.2 XP_008252886.1 11 - 12 2165 59
XM_008254665.2 XP_008252887.1 11 - 12 2165 59
XM_008254667.2 XP_008252889.1 11 - 12 2165 59
XM_008254670.2 XP_008252892.1 10 - 11 1805 59
XM_008254672.2 XP_008252894.1 9 - 10 1650 59
XM_008254673.2 XP_008252895.1 8 - 9 1580 59
XM_017340886.1 XP_017196375.1 8 - 9 1598 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0