This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06710711_g1
See other PRSS38 GE Assays ›
Gene Symbol:
PRSS38
Gene Name:
serine protease 38
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 46884 - 55783 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06710711_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS38
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008252778.2 XP_008251000.1 1327 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus

Panther Classification:

Molecular Function -

serine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0