Assay Details
| Gene Symbol: LOC100350827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262483.2 | XP_008260705.1 | 13 - 14 | 1755 | 69 | |
| XM_008262484.2 | XP_008260706.1 | 11 - 12 | 1649 | 69 | ||
| XM_017344959.1 | XP_017200448.1 | 14 - 15 | 1859 | 69 | ||
More Information
Set Membership: |
|

LOC100350827
Genomic Map