This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06721523_m1
See other NUDT9 GE Assays ›
Gene Symbol:
NUDT9
Gene Name:
nudix hydrolase 9
Gene Aliases:
-
Chromosome Location:
Chr.15: 62702538 - 62756864 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06721523_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002717030.3 XP_002717076.1 3 - 4 455 77
XM_008267620.2 XP_008265842.1 3 - 4 619 77
XM_008267621.2 XP_008265843.1 4 - 5 550 77
XM_008267622.2 XP_008265844.1 3 - 4 356 77
XM_008267623.2 XP_008265845.1 3 - 4 525 77
XM_017347419.1 XP_017202908.1 4 - 5 500 77
XM_017347420.1 XP_017202909.1 3 - 4 619 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0