This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06732987_m1
See other PPP1R11 GE Assays ›
Gene Symbol:
PPP1R11
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 11
Gene Aliases:
-
Chromosome Location:
Chr.12: 22060702 - 22064144 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06732987_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002714343.3 XP_002714389.1 2 - 3 330 71
XM_008262647.2 XP_008260869.2 3 - 4 205 71
XM_008262648.2 XP_008260870.1 3 - 4 233 71
XM_008262649.2 XP_008260871.1 3 - 4 201 71
XM_008262650.2 XP_008260872.1 2 - 3 193 71
XM_008262651.2 XP_008260873.1 3 - 4 157 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0