This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06739055_m1
See other CHST15 GE Assays ›
Gene Symbol:
CHST15
Gene Name:
carbohydrate sulfotransferase 15
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 1038908 - 1112225 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06739055_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHST15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008249390.2 XP_008247612.1 2328 61
XM_017338088.1 XP_017193577.1 2462 61
XM_017338089.1 XP_017193578.1 2198 61
XM_017338090.1 XP_017193579.1 2500 61
XM_017338091.1 XP_017193580.1 2190 61
XM_017338092.1 XP_017193581.1 2124 61
XM_017338093.1 XP_017193582.1 2127 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus

Panther Classification:

Molecular Function -

transferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline