This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06744715_m1
See other ARHGAP30 GE Assays ›
Gene Symbol:
ARHGAP30
Gene Name:
Rho GTPase activating protein 30
Gene Aliases:
-
Chromosome Location:
Chr.13: 31916517 - 31935745 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06744715_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP30
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002715255.3 XP_002715301.1 3 - 4 348 61
XM_017345747.1 XP_017201236.1 3 - 4 638 61
XM_017345748.1 XP_017201237.1 3 - 4 348 61
XM_017345749.1 XP_017201238.1 3 - 4 381 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017345749.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0