This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06744841_m1
See other LIG4 GE Assays ›
Gene Symbol:
LIG4
Gene Name:
DNA ligase 4
Gene Aliases:
-
Chromosome Location:
Chr.8: 109975508 - 109983237 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06744841_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIG4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008260173.2 XP_008258395.1 2 - 3 252 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008260173.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0