This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06755520_g1
See other ATXN1 GE Assays ›
Gene Symbol:
ATXN1
Gene Name:
ataxin 1
Gene Aliases:
sca1
Chromosome Location:
Chr.Unknown: 3258996 - 3709304 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06755520_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATXN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002720866.3 XP_002720912.1 403 79
XM_008274135.2 XP_008272357.1 536 79
XM_008274137.2 XP_008272359.1 598 79
XM_008274138.2 XP_008272360.1 465 79
XM_017350442.1 XP_017205931.1 365 79
XM_017350444.1 XP_017205933.1 403 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_002720866.3, XM_008274135.2, XM_008274137.2, XM_008274138.2, XM_017350442.1, XM_017350444.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Oryctolagus cuniculus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline