This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06759023_m1
See other AASDH GE Assays ›
Gene Symbol:
AASDH
Gene Name:
aminoadipate-semialdehyde dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.15: 92510644 - 92551062 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06759023_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AASDH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002717077.3 XP_002717123.2 6 - 7 1528 66
XM_008267824.2 XP_008266046.1 6 - 7 1528 66
XM_008267825.2 XP_008266047.1 6 - 7 1412 66
XM_008267827.2 XP_008266049.1 6 - 7 1355 66
XM_008267829.2 XP_008266051.1 5 - 6 1224 66
XM_008267832.2 XP_008266054.1 4 - 5 642 66
XM_017347539.1 XP_017203028.1 6 - 7 1589 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

ligase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0