This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06764594_m1
See other RAET1E GE Assays ›
Gene Symbol:
RAET1E
Gene Name:
retinoic acid early transcript 1E
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 30543 - 43449 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06764594_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAET1E
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008249898.2 XP_008248120.1 215 67
XM_017338347.1 XP_017193836.1 221 67
XM_017338348.1 XP_017193837.1 186 67
XM_017338350.1 XP_017193839.1 336 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008249898.2, XM_017338347.1, XM_017338348.1, XM_017338350.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Oryctolagus cuniculus

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0