Assay Details
| Gene Symbol: LOC100343496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269863.2 | XP_008268085.1 | 16 - 17 | 1654 | 93 | |
| XM_008269864.2 | XP_008268086.1 | 16 - 17 | 1651 | 93 | ||
| XM_008269865.2 | XP_008268087.1 | 16 - 17 | 1654 | 93 | ||
| XM_008269866.2 | XP_008268088.1 | 16 - 17 | 1654 | 93 | ||
| XM_008269867.2 | XP_008268089.1 | 16 - 17 | 1654 | 93 | ||
| XM_017348677.1 | XP_017204166.1 | 15 - 16 | 1546 | 93 | ||
More Information
Set Membership: |
|

LOC100343496
Genomic Map