This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06769708_m1
See other TMEM218 GE Assays ›
Gene Symbol:
TMEM218
Gene Name:
transmembrane protein 218
Gene Aliases:
-
Chromosome Location:
Chr.1: 92155211 - 92168138 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06769708_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM218
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002708703.3 XP_002708749.3 3 - 4 322 77
XM_017342739.1 XP_017198228.1 3 - 4 351 77
XM_017342745.1 XP_017198234.1 3 - 4 271 77
XM_017342749.1 XP_017198238.1 3 - 4 355 77
XM_017342754.1 XP_017198243.1 3 - 4 332 77
XM_017342755.1 XP_017198244.1 3 - 4 335 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0