This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06775976_m1
See other GPR1 GE Assays ›
Gene Symbol:
GPR1
Gene Name:
G protein-coupled receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 146953673 - 147008408 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06775976_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008259021.2 XP_008257243.1 3 - 4 387 80
XM_008259026.2 XP_008257248.1 5 - 6 531 80
XM_008259027.2 XP_008257249.1 3 - 4 668 80
XM_008259029.2 XP_008257251.1 4 - 5 423 80
XM_008259031.2 XP_008257253.1 2 - 3 177 80
XM_017343059.1 XP_017198548.1 4 - 5 495 80
XM_017343060.1 XP_017198549.1 2 - 3 566 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008259021.2, XM_008259026.2, XM_008259027.2, XM_008259029.2, XM_008259031.2, XM_017343059.1, XM_017343060.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline