This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06777141_m1
See other DSEL GE Assays ›
Gene Symbol:
DSEL
Gene Name:
dermatan sulfate epimerase like
Gene Aliases:
-
Chromosome Location:
Chr.9: 107629336 - 107635460 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06777141_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DSEL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008261447.2 XP_008259669.1 1 - 2 144 110
XM_017344226.1 XP_017199715.1 1 - 2 144 110

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008261447.2, XM_017344226.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0