Assay Details
| Gene Symbol: LOC100345689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263532.2 | XP_008261754.1 | 2 - 3 | 337 | 86 | |
| XM_017345379.1 | XP_017200868.1 | 2 - 3 | 340 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008263532.2, XM_017345379.1 transcripts.
Set Membership: |
|

LOC100345689
Genomic Map