This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06789678_m1
See other DISP1 GE Assays ›
Gene Symbol:
DISP1
Gene Name:
dispatched RND transporter family member 1
Gene Aliases:
-
Chromosome Location:
Chr.16: 50043249 - 50237507 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06789678_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DISP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002717490.3 XP_002717536.1 4 - 5 962 62
XM_008268371.2 XP_008266593.1 7 - 8 1043 62
XM_008268372.2 XP_008266594.1 6 - 7 966 62
XM_008268373.2 XP_008266595.1 6 - 7 945 62
XM_008268374.2 XP_008266596.1 5 - 6 868 62
XM_008268375.2 XP_008266597.1 7 - 8 1100 62
XM_008268377.2 XP_008266599.1 6 - 7 981 62
XM_008268378.2 XP_008266600.1 5 - 6 883 62
XM_008268379.2 XP_008266601.1 4 - 5 806 62
XM_008268383.2 XP_008266605.1 6 - 7 604 62
XM_008268384.2 XP_008266606.1 5 - 6 506 62
XM_008268385.2 XP_008266607.1 4 - 5 600 62
XM_008268386.2 XP_008266608.1 4 - 5 444 62
XM_017347770.1 XP_017203259.1 6 - 7 1002 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9w5rb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0