Assay Details
| Gene Symbol: LOC100341774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257389.2 | XP_008255611.1 | 1 - 2 | 183 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100341774| Gene Symbol: LOC100341774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257389.2 | XP_008255611.1 | 1 - 2 | 183 | 89 | |
Set Membership: |
|