This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06800735_m1
See other ETFBKMT GE Assays ›
Gene Symbol:
ETFBKMT
Gene Name:
electron transfer flavoprotein subunit beta lysine methyltransferase
Gene Aliases:
METTL20
Chromosome Location:
Chr.8: 9480018 - 9486347 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06800735_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ETFBKMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008259433.2 XP_008257655.1 1 - 2 147 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008259433.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline