This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03470948_g1
See other LOC4325486 GE Assays ›
Gene Symbol:
LOC4325486
Gene Name:
probable zinc metalloprotease EGY2, chloroplastic
Gene Aliases:
EGY2, OSNPB_010142100, OsEGY2, OsJ_00322
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03470948_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4325486
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015757621.2 XP_015613107.1 1 - 2 209 69
XM_015757626.2 XP_015613112.1 1 - 2 208 69
XM_015757635.2 XP_015613121.1 1 - 2 192 69
XM_015757642.2 XP_015613128.1 1 - 2 211 69
GenBank mRNA AK065031.1 - 1 - 2 206 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015757626.2, XM_015757635.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

metalloprotease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0