Assay Details
| Gene Symbol: LOC4328814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767665.1 | XP_015623151.1 | 3 - 4 | 823 | 63 | |
| GenBank mRNA | AB856419.1 | - | 3 - 4 | 628 | 63 | |
| AK068806.1 | - | 3 - 4 | 724 | 63 | ||
More Information
Set Membership: |
|

LOC4328814
Genomic Map