Assay Details
| Gene Symbol: LOC4336345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781188.2 | XP_015636674.1 | 12 - 13 | 1438 | 128 | |
| XM_015781189.2 | XP_015636675.1 | 12 - 13 | 1372 | 128 | ||
| GenBank mRNA | AK067042.1 | - | 12 - 13 | 1425 | 128 | |
More Information
Set Membership: |
|

LOC4336345
Genomic Map