This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03499918_g1
See other LOC4345304 GE Assays ›
Gene Symbol:
LOC4345304
Gene Name:
pre-mRNA-splicing factor ATP-dependent RNA helicase DEAH1
Gene Aliases:
OSNPB_080335000
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03499918_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4345304
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015794949.2 XP_015650435.1 18 - 19 1702 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

RNA helicase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline