This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03503879_m1
See other LOC4326932 GE Assays ›
Gene Symbol:
LOC4326932
Gene Name:
bifunctional nuclease 1-like
Gene Aliases:
BBD1, OSNPB_010702000, OsBBD1, OsJ_03158, P0421H07.6
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03503879_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4326932
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015765834.2 XP_015621320.1 6 - 7 824 108
XM_015765836.2 XP_015621322.1 6 - 7 819 108
XM_015765837.2 XP_015621323.1 6 - 7 824 108
XM_015765838.2 XP_015621324.1 6 - 7 824 108
GenBank mRNA AK065414.1 - 6 - 7 777 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline