Assay Details
| Gene Symbol: LOC4338710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784981.2 | XP_015640467.2 | 1 - 2 | 452 | 150 | |
| GenBank mRNA | AK102894.1 | - | 1 - 2 | 206 | 150 | |
More Information
Set Membership: |
|

LOC4338710
Genomic Map