Assay Details
| Gene Symbol: LOC4342538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791204.2 | XP_015646690.1 | 8 - 9 | 1945 | 75 | |
| GenBank mRNA | AK062003.1 | - | 5 - 6 | 591 | 75 | |
| AK065897.1 | - | 8 - 9 | 1947 | 75 | ||
More Information
Set Membership: |
|

LOC4342538
Genomic Map