This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03534053_g1
See other LOC4330159 GE Assays ›
Gene Symbol:
LOC4330159
Gene Name:
ATP-dependent zinc metalloprotease FTSH 7, chloroplastic-like
Gene Aliases:
FTSH7, OSNPB_020649700, OsFTSH7, OsJ_007487
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03534053_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4330159
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015769923.2 XP_015625409.1 7 - 8 1736 88
GenBank mRNA AK058221.1 - 7 - 8 813 88
AK069509.1 - 7 - 8 1739 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

metalloprotease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline