Assay Details
| Gene Symbol: LOC9267485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782797.2 | XP_015638283.1 | 15 - 16 | 1342 | 114 | |
| XM_015782799.2 | XP_015638285.1 | 15 - 16 | 1322 | 114 | ||
| GenBank mRNA | AK073973.1 | - | 10 - 11 | 2281 | 114 | |
More Information
Set Membership: |
|

LOC9267485
Genomic Map