Assay Details
| Gene Symbol: LOC9267485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782797.2 | XP_015638283.1 | 1 - 2 | 93 | 68 | |
| GenBank mRNA | AK073973.1 | - | 1 - 2 | 94 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015782797.2 transcripts.
Set Membership: |
|

LOC9267485
Genomic Map