Assay Details
| Gene Symbol: LOC4350166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759693.2 | XP_015615179.1 | 7 - 8 | 938 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4350166| Gene Symbol: LOC4350166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759693.2 | XP_015615179.1 | 7 - 8 | 938 | 76 | |
Set Membership: |
|