Assay Details
| Gene Symbol: LOC4327327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK101742.1 | - | 1 | 1671 | 144 | |
| U25430.1 | - | 1 | 1649 | 144 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4327327| Gene Symbol: LOC4327327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK101742.1 | - | 1 | 1671 | 144 | |
| U25430.1 | - | 1 | 1649 | 144 | ||
Set Membership: |
|