This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03593377_m1
See other LOC4343851 GE Assays ›
Gene Symbol:
LOC4343851
Gene Name:
MADS-box transcription factor 18-like
Gene Aliases:
FDRMADS7, MADS18, MADS2, MADS28, OSNPB_070605200, OsMADS18, OsMADS2, OsMADS28
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03593377_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4343851
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015791998.2 XP_015647484.1 1 - 2 292 80
GenBank mRNA AJ011675.1 - 1 - 2 259 80
AK064704.1 - 1 - 2 272 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

MADS box transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0