This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03595937_m1
See other LOC4325567 GE Assays ›
Gene Symbol:
LOC4325567
Gene Name:
probable pre-mRNA-splicing factor ATP-dependent RNA helicase DEAH9
Gene Aliases:
OSNPB_010212100
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03595937_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4325567
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015766618.2 XP_015622104.1 2 - 3 342 63
XM_015766619.2 XP_015622105.1 2 - 3 343 63
GenBank mRNA AK065467.1 - 2 - 3 302 63
AK101456.1 - 2 - 3 338 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

RNA helicase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline