This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03603327_m1
See other LOC4330484 GE Assays ›
Gene Symbol:
LOC4330484
Gene Name:
calpain-type cysteine protease ADL1-like
Gene Aliases:
49D11.1, ADL1, DEK1, ODM63, OJ1311_H06.4, OSNPB_020709400, OsDEK1, SHL3
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03603327_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4330484
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015769939.2 XP_015625425.1 29 - 30 6265 118
XM_015769940.2 XP_015625426.1 28 - 29 6028 118
GenBank mRNA AB477099.1 - 28 - 29 5951 118
AK064381.1 - 10 - 11 2376 118
AK072218.1 - 3 - 4 550 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

cysteine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0