This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03603336_g1
See other LOC4330484 GE Assays ›
Gene Symbol:
LOC4330484
Gene Name:
calpain-type cysteine protease ADL1-like
Gene Aliases:
49D11.1, ADL1, DEK1, ODM63, OJ1311_H06.4, OSNPB_020709400, OsDEK1, SHL3
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03603336_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4330484
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015769939.2 XP_015625425.1 9 - 10 2405 76
XM_015769940.2 XP_015625426.1 8 - 9 2168 76
GenBank mRNA AB477099.1 - 8 - 9 2091 76
AK121261.1 - 9 - 10 2405 76
AK121875.1 - 9 - 10 2360 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

cysteine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0